Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAACTTCCGACCTCAGGTGATCTGC[C/T]TGCCTCAGCCTCCCAAAGTGCTGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609884 | ||||||||||||||||||||
Literature Links: |
TMEM67 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TMEM67 - transmembrane protein 67 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142301.1 | 309 | Silent Mutation | CTG,TTG | L,L 22 | NP_001135773.1 | |
NM_153704.5 | 309 | Intron | NP_714915.3 | |||
XM_006716686.3 | 309 | Intron | XP_006716749.1 | |||
XM_011517363.2 | 309 | Intron | XP_011515665.1 |