Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTAGCAAGAGAGAAAGCTGTGCCG[A/G]GAGTGGTTGGGAAAGAGGGAGGGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613895 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DPY19L4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DPY19L4 - dpy-19 like 4 (C. elegans) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_181787.2 | Intron | NP_861452.2 | ||||
XM_005250894.4 | Intron | XP_005250951.2 | ||||
XM_005250895.3 | Intron | XP_005250952.1 | ||||
XM_011516983.2 | Intron | XP_011515285.1 | ||||
XM_011516984.2 | Intron | XP_011515286.1 | ||||
XM_017013351.1 | Intron | XP_016868840.1 | ||||
XM_017013352.1 | Intron | XP_016868841.1 | ||||
XM_017013353.1 | Intron | XP_016868842.1 | ||||
XM_017013354.1 | Intron | XP_016868843.1 | ||||
XM_017013355.1 | Intron | XP_016868844.1 | ||||
XM_017013356.1 | Intron | XP_016868845.1 | ||||
XM_017013357.1 | Intron | XP_016868846.1 |