Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGCCTCGGGCGGCCTTTTGAGGGG[C/T]TCGGCTAGCTTCAGCGCCGTGGGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609170 MIM: 243305 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ERP44 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ERP44 - endoplasmic reticulum protein 44 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015051.2 | 102 | Intron | NP_055866.1 |
INVS - inversin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318381.1 | 102 | UTR 5 | NP_001305310.1 | |||
NM_001318382.1 | 102 | UTR 5 | NP_001305311.1 | |||
NM_014425.4 | 102 | UTR 5 | NP_055240.2 |