Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCTACAATGAAGAGGAATTGCCC[C/T]TGGGCCTCTTCAATAGGAAACAAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602938 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BAAT PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BAAT - bile acid-CoA:amino acid N-acyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127610.1 | 1161 | Silent Mutation | CAA,CAG | Q,Q 317 | NP_001121082.1 | |
NM_001701.3 | 1161 | Silent Mutation | CAA,CAG | Q,Q 317 | NP_001692.1 |