Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTGTAGCCAACTTCCTGCAGGCCC[A/G]GGGCCTGGCCTCGGGCGATGTGGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604194 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC27A4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC27A4 - solute carrier family 27 member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005094.3 | 520 | Missense Mutation | CAG,CGG | Q,R 121 | NP_005085.2 | |
XM_017014222.1 | 520 | Missense Mutation | CAG,CGG | Q,R 121 | XP_016869711.1 |