Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTAATGCAATTTAATATCCCTAAT[A/G]CTTATAAAACACTTTGTATATCCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 120325 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COL15A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COL15A1 - collagen type XV alpha 1 chain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001855.4 | Intron | NP_001846.3 | ||||
XM_011518214.2 | Intron | XP_011516516.1 |