Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGTTTCAGACAGTTTCTTCAGAT[A/C]GGAGCTGTAATTCACCCAAAGTTTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605506 | ||||||||||||||||||||
Literature Links: |
VPS26A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
VPS26A - VPS26, retromer complex component A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001035260.2 | Intron | NP_001030337.1 | ||||
NM_001318944.1 | Intron | NP_001305873.1 | ||||
NM_001318945.1 | Intron | NP_001305874.1 | ||||
NM_001318946.1 | Intron | NP_001305875.1 | ||||
NM_004896.4 | Intron | NP_004887.2 |