Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGGAAGAAAAAGTTGGTGAAGAA[C/T]ATTTAGGACTCTCACACTAAATACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 194528 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF25 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF25 - zinc finger protein 25 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005252384.2 | 2975 | UTR 3 | XP_005252441.1 | |||
XM_005252385.2 | 2975 | UTR 3 | XP_005252442.1 | |||
XM_005252386.2 | 2975 | UTR 3 | XP_005252443.1 | |||
XM_011519352.2 | 2975 | UTR 3 | XP_011517654.1 | |||
XM_011519353.2 | 2975 | UTR 3 | XP_011517655.1 | |||
XM_011519354.2 | 2975 | UTR 3 | XP_011517656.1 | |||
XM_011519355.2 | 2975 | UTR 3 | XP_011517657.1 | |||
XM_011519356.2 | 2975 | UTR 3 | XP_011517658.1 | |||
XM_017015837.1 | 2975 | UTR 3 | XP_016871326.1 | |||
XM_017015838.1 | 2975 | UTR 3 | XP_016871327.1 |