Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACATCATCTTGTGATAACAGATCA[A/G]TTTTAATTCTAGCACCTGAAGCTAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600098 MIM: 604989 | ||||||||||||||||||||
Literature Links: |
RRAS2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RRAS2 - related RAS viral (r-ras) oncogene homolog 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001102669.2 | 2177 | UTR 3 | NP_001096139.1 | |||
NM_001177314.1 | 2177 | Intron | NP_001170785.1 | |||
NM_001177315.1 | 2177 | Intron | NP_001170786.1 | |||
NM_012250.5 | 2177 | UTR 3 | NP_036382.2 | |||
XM_017017363.1 | 2177 | Intron | XP_016872852.1 |
SPON1 - spondin 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |