Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGTGTCTCCTTTAAGACTTTCAA[A/G]ATATTAAGATTCAAAAGTTACGAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607257 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR6073 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR6073 - microRNA 6073 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SOX6 - SRY-box 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145811.1 | 3191 | UTR 3 | NP_001139283.1 | |||
NM_001145819.1 | 3191 | UTR 3 | NP_001139291.1 | |||
NM_017508.2 | 3191 | UTR 3 | NP_059978.1 | |||
NM_033326.3 | 3191 | UTR 3 | NP_201583.2 |