Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGTCATAGTTGCTTTGAACTGTG[C/T]AATGAGTAATAAACTTGGGTTGAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610110 MIM: 612455 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANO3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANO3 - anoctamin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001313726.1 | 5621 | Intron | NP_001300655.1 | |||
NM_001313727.1 | 5621 | Intron | NP_001300656.1 | |||
NM_031418.3 | 5621 | Intron | NP_113606.2 |
SLC5A12 - solute carrier family 5 member 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178498.3 | 5621 | UTR 3 | NP_848593.2 | |||
XM_006718155.3 | 5621 | UTR 3 | XP_006718218.1 | |||
XM_006718156.3 | 5621 | Intron | XP_006718219.1 | |||
XM_011519920.2 | 5621 | UTR 3 | XP_011518222.1 | |||
XM_011519921.2 | 5621 | Intron | XP_011518223.1 | |||
XM_017017244.1 | 5621 | UTR 3 | XP_016872733.1 |