Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAACCTGCAGTATTTTCAACCCT[A/C]ACTCCTTGTTGCTTTGTGCAATGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
QSER1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
QSER1 - glutamine and serine rich 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001076786.2 | Intron | NP_001070254.1 | ||||
XM_006718325.3 | Intron | XP_006718388.3 | ||||
XM_017018327.1 | Intron | XP_016873816.1 | ||||
XM_017018328.1 | Intron | XP_016873817.1 | ||||
XM_017018329.1 | Intron | XP_016873818.1 | ||||
XM_017018330.1 | Intron | XP_016873819.1 |