Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGGTGGCCCAGAACTTTTGAGTGC[A/G]CACTTTGTTGGCTGATCACAGGTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610509 MIM: 601197 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RIC3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RIC3 - RIC3 acetylcholine receptor chaperone | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135109.2 | Intron | NP_001128581.1 | ||||
NM_001206671.2 | Intron | NP_001193600.1 | ||||
NM_001206672.2 | Intron | NP_001193601.1 | ||||
NM_024557.4 | Intron | NP_078833.3 | ||||
XM_006718317.3 | Intron | XP_006718380.1 | ||||
XM_006718318.3 | Intron | XP_006718381.1 | ||||
XM_006718321.3 | Intron | XP_006718384.1 | ||||
XM_017018287.1 | Intron | XP_016873776.1 | ||||
XM_017018288.1 | Intron | XP_016873777.1 | ||||
XM_017018289.1 | Intron | XP_016873778.1 | ||||
XM_017018290.1 | Intron | XP_016873779.1 |
TUB - tubby bipartite transcription factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003320.4 | Intron | NP_003311.2 | ||||
NM_177972.2 | Intron | NP_813977.1 | ||||
XM_005253109.3 | Intron | XP_005253166.1 | ||||
XM_011520344.2 | Intron | XP_011518646.1 |