Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAGATTGTGACAAGCTTTTCCTTT[A/C]CTTAAAAAAAGAAAATACTTAAAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
DENND5A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DENND5A - DENN domain containing 5A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243254.1 | 3176 | UTR 3 | NP_001230183.1 | |||
NM_015213.3 | 3176 | UTR 3 | NP_056028.2 | |||
XM_005252832.1 | 3176 | UTR 3 | XP_005252889.1 | |||
XM_011519953.2 | 3176 | UTR 3 | XP_011518255.1 | |||
XM_017017419.1 | 3176 | Intron | XP_016872908.1 |