Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCACAATTCTAAGAGGAAAGCACTC[C/T]TTCTTTTTTTCTTTTTTTTTTTTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606264 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLEC7A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CLEC7A - C-type lectin domain family 7 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022570.4 | Intron | NP_072092.2 | ||||
NM_197947.2 | Intron | NP_922938.1 | ||||
NM_197948.2 | Intron | NP_922939.1 | ||||
NM_197949.2 | Intron | NP_922940.1 | ||||
NM_197950.2 | Intron | NP_922941.1 | ||||
NM_197954.2 | Intron | NP_922945.1 | ||||
XM_006719135.3 | Intron | XP_006719198.1 | ||||
XM_017019822.1 | Intron | XP_016875311.1 | ||||
XM_017019823.1 | Intron | XP_016875312.1 |