Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGCTGCTAACAAATGATGATACC[A/G]GAAGTGGGTACCTGAGTGTCGGTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607363 | ||||||||||||||||||||
Literature Links: |
KNTC1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KNTC1 - kinetochore associated 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014708.4 | 185 | Missense Mutation | AGA,GGA | R,G 14 | NP_055523.1 | |
XM_006719706.2 | 185 | Missense Mutation | AGA,GGA | R,G 14 | XP_006719769.1 | |
XM_011539027.2 | 185 | Missense Mutation | AGA,GGA | R,G 14 | XP_011537329.1 | |
XM_011539029.2 | 185 | Missense Mutation | AGA,GGA | R,G 14 | XP_011537331.1 | |
XM_011539030.1 | 185 | Missense Mutation | AGA,GGA | R,G 14 | XP_011537332.1 | |
XM_017020254.1 | 185 | Missense Mutation | AGA,GGA | R,G 14 | XP_016875743.1 | |
XM_017020255.1 | 185 | Intron | XP_016875744.1 | |||
XM_017020256.1 | 185 | Intron | XP_016875745.1 | |||
XM_017020257.1 | 185 | Missense Mutation | AGA,GGA | R,G 14 | XP_016875746.1 |
RSRC2 - arginine and serine rich coiled-coil 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |