Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAACCAGCTGTCTCCATATGAACA[A/T]AGTGAAATCCTGGGCTACGCGGAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609181 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DYRK4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DYRK4 - dual specificity tyrosine phosphorylation regulated kinase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282285.1 | 328 | Intron | NP_001269214.1 | |||
NM_001282286.1 | 328 | Intron | NP_001269215.1 | |||
NM_003845.2 | 328 | Missense Mutation | CAA,CAT | Q,H 54 | NP_003836.1 |