Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCACGCCACCCGGCCCGCCGCCCCC[C/T]GACACCAACAGCATGGAGCTGGGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611055 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LINC01089 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LINC01089 - long intergenic non-protein coding RNA 1089 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SETD1B - SET domain containing 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015048.1 | 1536 | Silent Mutation | CCC,CCT | P,P 457 | NP_055863.1 | |
XM_005253858.4 | 1536 | Silent Mutation | CCC,CCT | P,P 457 | XP_005253915.1 | |
XM_006719296.3 | 1536 | Silent Mutation | CCC,CCT | P,P 457 | XP_006719359.1 |