Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGGAAAACGGGCGAGGAGGAAGG[A/G]GAGGAGGAGGGGAAGGAGGACAAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603146 | ||||||||||||||||||||
Literature Links: |
PSMD9 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PSMD9 - proteasome 26S subunit, non-ATPase 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WDR66 - WD repeat domain 66 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001178003.1 | 358 | Silent Mutation | GGA,GGG | G,G 62 | NP_001171474.1 | |
NM_144668.5 | 358 | Silent Mutation | GGA,GGG | G,G 62 | NP_653269.3 |