Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTCTATTTTTCCTAATTGTGTTC[A/C]TATTTTTATTTAAGTCCTTGAACAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605002 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CORO2B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CORO2B - coronin 2B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001190456.1 | Intron | NP_001177385.1 | ||||
NM_001190457.1 | Intron | NP_001177386.1 | ||||
NM_001324014.1 | Intron | NP_001310943.1 | ||||
NM_001324015.1 | Intron | NP_001310944.1 | ||||
NM_006091.4 | Intron | NP_006082.3 |