Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTATCCAGACTATCCACATTCTCCA[A/T]GGTCTTCTTGCTGTTTCCTTCCTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 154580 MIM: 607776 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAN2C1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAN2C1 - mannosidase alpha class 2C member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SIN3A - SIN3 transcription regulator family member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145357.1 | 3648 | Missense Mutation | ATG,TTG | M,L 1156 | NP_001138829.1 | |
NM_001145358.1 | 3648 | Missense Mutation | ATG,TTG | M,L 1156 | NP_001138830.1 | |
NM_015477.2 | 3648 | Missense Mutation | ATG,TTG | M,L 1156 | NP_056292.1 | |
XM_006720465.3 | 3648 | Missense Mutation | ATG,TTG | M,L 1156 | XP_006720528.1 | |
XM_006720466.3 | 3648 | Missense Mutation | ATG,TTG | M,L 1156 | XP_006720529.1 | |
XM_006720467.3 | 3648 | Missense Mutation | ATG,TTG | M,L 1156 | XP_006720530.1 |