Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTACACCATCTTTGGTCTATATCAA[A/T]CAAAGGAAAACAAGAATAGTGAAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616142 MIM: 603962 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM98B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM98B - family with sequence similarity 98 member B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RASGRP1 - RAS guanyl releasing protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128602.1 | 4217 | UTR 3 | NP_001122074.1 | |||
NM_001306086.1 | 4217 | UTR 3 | NP_001293015.1 | |||
NM_005739.3 | 4217 | UTR 3 | NP_005730.2 | |||
XM_005254114.2 | 4217 | UTR 3 | XP_005254171.1 | |||
XM_011521151.2 | 4217 | UTR 3 | XP_011519453.1 | |||
XM_017021860.1 | 4217 | UTR 3 | XP_016877349.1 |