Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACACTCAGAGGCTTTATTATCCAC[A/C]CCTACCCCCGCCCCACCTACCCTAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 191845 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PDILT PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PDILT - protein disulfide isomerase like, testis expressed | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_174924.1 | 1282 | UTR 3 | NP_777584.1 | |||
XM_011545764.1 | 1282 | Intron | XP_011544066.1 | |||
XM_011545765.1 | 1282 | Intron | XP_011544067.1 | |||
XM_011545766.2 | 1282 | UTR 3 | XP_011544068.1 |
UMOD - uromodulin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |