Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTGAAGATATAAAAAATTAAAAT[C/T]TATAGAAAATGGCAGGTTATTTTTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
USP31 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
USP31 - ubiquitin specific peptidase 31 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020718.3 | 7757 | UTR 3 | NP_065769.3 | |||
XM_017023496.1 | 7757 | UTR 3 | XP_016878985.1 | |||
XM_017023497.1 | 7757 | UTR 3 | XP_016878986.1 | |||
XM_017023498.1 | 7757 | UTR 3 | XP_016878987.1 | |||
XM_017023499.1 | 7757 | UTR 3 | XP_016878988.1 | |||
XM_017023500.1 | 7757 | Intron | XP_016878989.1 |