Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCACTGCGAACTTCCTCTTGAAG[A/C]GCATGCCACACTCGGGGCAGGGGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF688 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF688 - zinc finger protein 688 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001024683.1 | 746 | Missense Mutation | CGC,CTC | R,L 205 | NP_001019854.1 | |
NM_145271.3 | 746 | Missense Mutation | CGC,CTC | R,L 219 | NP_660314.1 | |
XM_005255139.2 | 746 | Missense Mutation | CGC,CTC | R,L 142 | XP_005255196.1 | |
XM_005255140.2 | 746 | Missense Mutation | CGC,CTC | R,L 142 | XP_005255197.1 |
ZNF785 - zinc finger protein 785 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |