Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 158343 | ||||||||||||||||||||
Literature Links: |
ABCC1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ABCC1 - ATP binding cassette subfamily C member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004996.3 | 202 | Silent Mutation | GCC,GCG | A,A 9 | NP_004987.2 | |
XM_011522497.1 | 202 | Intron | XP_011520799.1 | |||
XM_011522498.2 | 202 | Intron | XP_011520800.1 | |||
XM_017023237.1 | 202 | Intron | XP_016878726.1 | |||
XM_017023238.1 | 202 | Intron | XP_016878727.1 | |||
XM_017023239.1 | 202 | Intron | XP_016878728.1 | |||
XM_017023240.1 | 202 | Intron | XP_016878729.1 | |||
XM_017023241.1 | 202 | Intron | XP_016878730.1 | |||
XM_017023242.1 | 202 | Intron | XP_016878731.1 | |||
XM_017023243.1 | 202 | Intron | XP_016878732.1 |