Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCAAGTCATCAATTAGTCACAGAC[C/T]CAGAATTTGTAGATTCCAAGAAACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602212 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100507577 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC100507577 - uncharacterized LOC100507577 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SIAH1 - siah E3 ubiquitin protein ligase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001006610.1 | Intron | NP_001006611.1 | ||||
NM_003031.3 | Intron | NP_003022.3 | ||||
XM_006721246.1 | Intron | XP_006721309.1 | ||||
XM_011523279.1 | Intron | XP_011521581.1 |