Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCACCACGCCCGGCCCCTATTTT[A/G]CTCTTGTTTAGGTAATGTCATTCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604167 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CTCF PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CTCF - CCCTC-binding factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001191022.1 | Intron | NP_001177951.1 | ||||
NM_006565.3 | Intron | NP_006556.1 | ||||
XM_005255775.3 | Intron | XP_005255832.1 | ||||
XM_017022868.1 | Intron | XP_016878357.1 | ||||
XM_017022869.1 | Intron | XP_016878358.1 |