Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTATAAGGTGGCTGTGCCGCCTGC[A/C]ACCTTTTCACCAACTGGACTCCCAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614301 | ||||||||||||||||||||
Literature Links: |
ATXN1L PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ATXN1L - ataxin 1 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001137675.3 | 563 | Silent Mutation | GCA,GCC | A,A 90 | NP_001131147.1 |
ZNF821 - zinc finger protein 821 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |