Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTATGACAAGGACACCACCAAACC[A/T]ATCAGCTTTTACTTGTCTTCGCTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611898 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ENGASE PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ENGASE - endo-beta-N-acetylglucosaminidase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042573.2 | 96 | Silent Mutation | CCA,CCT | P,P 82 | NP_001036038.1 | |
XM_006722017.3 | 96 | Intron | XP_006722080.1 | |||
XM_006722018.3 | 96 | Intron | XP_006722081.1 | |||
XM_006722019.3 | 96 | Silent Mutation | CCA,CCT | P,P 82 | XP_006722082.1 | |
XM_017024961.1 | 96 | UTR 5 | XP_016880450.1 |