Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTTTTTACTCACTCCTGCTTCCT[C/G]CCAAATAATCCCTCACTGGGCTGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607217 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SDK2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SDK2 - sidekick cell adhesion molecule 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144952.1 | 9036 | UTR 3 | NP_001138424.1 | |||
XM_011524914.2 | 9036 | UTR 3 | XP_011523216.1 | |||
XM_011524915.2 | 9036 | UTR 3 | XP_011523217.1 | |||
XM_011524916.2 | 9036 | Intron | XP_011523218.1 |