Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAATAGTACTAGATAGTATGGTTG[G/T]CATGTTAGAGAATAGCATATTCTGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601335 | ||||||||||||||||||||
Literature Links: |
MAP2K4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MAP2K4 - mitogen-activated protein kinase kinase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281435.1 | Intron | NP_001268364.1 | ||||
NM_003010.3 | Intron | NP_003001.1 | ||||
XM_005256753.2 | Intron | XP_005256810.1 | ||||
XM_005256755.2 | Intron | XP_005256812.1 | ||||
XM_011523976.2 | Intron | XP_011522278.1 | ||||
XM_011523977.2 | Intron | XP_011522279.1 |