Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCATGGCCCATTTCAACACTGGA[C/T]GGCCCGTGGCGAACCTCCACCACGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 172860 | ||||||||||||||||||||
Literature Links: |
SERPINF1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SERPINF1 - serpin family F member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SMYD4 - SET and MYND domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_052928.2 | 2618 | Silent Mutation | CCA,CCG | P,P 735 | NP_443160.2 | |
XM_011523632.2 | 2618 | Intron | XP_011521934.1 | |||
XM_011523633.2 | 2618 | Intron | XP_011521935.1 | |||
XM_017024121.1 | 2618 | Silent Mutation | CCA,CCG | P,P 631 | XP_016879610.1 |