Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCACGAAAAAGATGACTGAAAGTT[C/T]AAAAGTCCTAAGTGTAGGCACTTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607927 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKFY1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANKFY1 - ankyrin repeat and FYVE domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005256679.3 | 6567 | UTR 3 | XP_005256736.1 | |||
XM_011523926.1 | 6567 | UTR 3 | XP_011522228.1 | |||
XM_011523928.2 | 6567 | UTR 3 | XP_011522230.1 | |||
XM_011523933.1 | 6567 | UTR 3 | XP_011522235.1 | |||
XM_017024733.1 | 6567 | UTR 3 | XP_016880222.1 |
CYB5D2 - cytochrome b5 domain containing 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |