Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCATACTCCCGCTTGGGCCGTGT[C/T]GTATCTGCCAGGAAGTCACCCCACC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602887 | ||||||||||||||||||||
Literature Links: |
DLG4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DLG4 - discs large MAGUK scaffold protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128827.2 | 1825 | Silent Mutation | ACC,ACT | T,T 563 | NP_001122299.1 | |
NM_001321074.1 | 1825 | Silent Mutation | ACC,ACT | T,T 606 | NP_001308003.1 | |
NM_001321075.1 | 1825 | Silent Mutation | ACC,ACT | T,T 566 | NP_001308004.1 | |
NM_001321076.1 | 1825 | Silent Mutation | ACC,ACT | T,T 506 | NP_001308005.1 | |
NM_001321077.1 | 1825 | Silent Mutation | ACC,ACT | T,T 506 | NP_001308006.1 | |
NM_001365.4 | 1825 | Silent Mutation | ACC,ACT | T,T 609 | NP_001356.1 | |
XM_005256491.1 | 1825 | Silent Mutation | XP_005256548.1 | |||
XM_011523698.1 | 1825 | Silent Mutation | XP_011522000.1 | |||
XM_011523699.2 | 1825 | Silent Mutation | XP_011522001.1 | |||
XM_011523702.1 | 1825 | Silent Mutation | XP_011522004.1 | |||
XM_017024288.1 | 1825 | Silent Mutation | XP_016879777.1 | |||
XM_017024289.1 | 1825 | Silent Mutation | XP_016879778.1 | |||
XM_017024290.1 | 1825 | Silent Mutation | XP_016879779.1 |