Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGCGGCGGGGTCCAGGGATGGGCG[C/T]GGCGCTGGAGGCCTCGGATCTACCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607206 MIM: 608059 MIM: 610407 | ||||||||||||||||||||
Literature Links: |
ALOXE3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ALOXE3 - arachidonate lipoxygenase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HES7 - hes family bHLH transcription factor 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001165967.1 | 2046 | Silent Mutation | CCA,CCG | P,P 148 | NP_001159439.1 | |
NM_032580.3 | 2046 | Silent Mutation | CCA,CCG | P,P 143 | NP_115969.2 | |
XM_011524041.2 | 2046 | Silent Mutation | CCA,CCG | P,P 177 | XP_011522343.1 | |
XM_017025232.1 | 2046 | Silent Mutation | CCA,CCG | P,P 183 | XP_016880721.1 |
TRG-GCC2-6 - transfer RNA-Gly (GCC) 2-6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |