Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTACCCATCCTTCTCCTTCTACTGC[C/G]AGGAATCTAAGAGGCTATGGGGTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611317 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PIK3R5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PIK3R5 - phosphoinositide-3-kinase regulatory subunit 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142633.2 | 2831 | UTR 3 | NP_001136105.1 | |||
NM_001251851.1 | 2831 | UTR 3 | NP_001238780.1 | |||
NM_001251852.1 | 2831 | UTR 3 | NP_001238781.1 | |||
NM_001251853.1 | 2831 | UTR 3 | NP_001238782.1 | |||
NM_001251855.1 | 2831 | UTR 3 | NP_001238784.1 | |||
NM_014308.3 | 2831 | UTR 3 | NP_055123.2 | |||
XM_005256580.3 | 2831 | UTR 3 | XP_005256637.1 |