Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAATATGGTAATTAAGACTAGGAA[G/T]GGAGCAGTTCAGTCTAAAAAATATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 164762 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CRK PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CRK - CRK proto-oncogene, adaptor protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005206.4 | 1216 | UTR 3 | NP_005197.3 | |||
NM_016823.3 | 1216 | UTR 3 | NP_058431.2 |