Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCCATGTTCCCTGCACTCCAGTT[A/C]CGACCCCAGAGCAGGAGGGAAGACG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 114207 | ||||||||||||||||||||
Literature Links: |
CACNB1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CACNB1 - calcium voltage-gated channel auxiliary subunit beta 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000723.4 | 1819 | Intron | NP_000714.3 | |||
NM_199247.2 | 1819 | UTR 3 | NP_954855.1 | |||
NM_199248.2 | 1819 | UTR 3 | NP_954856.1 | |||
XM_005257645.2 | 1819 | Intron | XP_005257702.1 | |||
XM_005257646.2 | 1819 | Intron | XP_005257703.1 | |||
XM_006722072.2 | 1819 | Intron | XP_006722135.1 | |||
XM_017025024.1 | 1819 | Intron | XP_016880513.1 | |||
XM_017025025.1 | 1819 | Intron | XP_016880514.1 | |||
XM_017025026.1 | 1819 | Intron | XP_016880515.1 | |||
XM_017025027.1 | 1819 | Intron | XP_016880516.1 | |||
XM_017025028.1 | 1819 | Intron | XP_016880517.1 | |||
XM_017025029.1 | 1819 | Intron | XP_016880518.1 | |||
XM_017025030.1 | 1819 | Intron | XP_016880519.1 |