Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCGCTCTCCAGCAGGCTCCGGTA[C/T]GTGTTGATCTCACACTCCAGCCGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602761 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KRT33A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KRT33A - keratin 33A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004138.3 | 869 | Silent Mutation | ACA,ACG | T,T 355 | NP_004129.2 | |
XM_011524786.2 | 869 | Silent Mutation | ACA,ACG | T,T 196 | XP_011523088.1 |