Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACTCACTGGGCATCCTCTCCCTCC[A/G]GCAGGCGGCGGTAGGTGGCAATCTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 148067 MIM: 148069 | ||||||||||||||||||||
Literature Links: |
KRT16 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KRT16 - keratin 16 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KRT17 - keratin 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000422.2 | Intron | NP_000413.1 |
KRT42P - keratin 42 pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |