Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCACAAGGCTTAATTTCTCAAATAA[A/G]TCTACTGAGCTTTTCTAATGGTTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603808 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MED13 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MED13 - mediator complex subunit 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005121.2 | 8468 | UTR 3 | NP_005112.2 | |||
XM_011525551.2 | 8468 | UTR 3 | XP_011523853.1 | |||
XM_011525552.2 | 8468 | Intron | XP_011523854.1 | |||
XM_011525553.2 | 8468 | UTR 3 | XP_011523855.1 |