Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGTGTGCACTGCAGGTCGATGCC[C/G]GAGTCTGGCACCTCCATGTCGTAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604655 | ||||||||||||||||||||
Literature Links: |
MAP3K14 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MAP3K14 - mitogen-activated protein kinase kinase kinase 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003954.4 | 3016 | Silent Mutation | TCC,TCG | S,S 918 | NP_003945.2 | |
XM_011525441.2 | 3016 | Silent Mutation | TCC,TCG | S,S 918 | XP_011523743.1 |
MAP3K14-AS1 - MAP3K14 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPATA32 - spermatogenesis associated 32 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |