Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATACAATTCTTACTTCGAATAAA[A/G]TTGTTCCAATGAGGACTGCATAGTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604983 | ||||||||||||||||||||
Literature Links: |
MILR1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MILR1 - mast cell immunoglobulin like receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001085423.1 | 1380 | Intron | NP_001078892.1 | |||
NM_001291316.1 | 1380 | Intron | NP_001278245.1 | |||
NM_001291317.1 | 1380 | Intron | NP_001278246.1 |
POLG2 - DNA polymerase gamma 2, accessory subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007215.3 | 1380 | Missense Mutation | CTT,TTT | L,F 428 | NP_009146.2 | |
XM_006721651.2 | 1380 | Intron | XP_006721714.1 | |||
XM_017024072.1 | 1380 | Intron | XP_016879561.1 |