Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGATTGTTTCAAAAATGTTCAGAGA[C/T]TCTGTAAGGCGATTGGTCAGTCGAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604406 | ||||||||||||||||||||
Literature Links: |
GNA13 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GNA13 - G protein subunit alpha 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282425.1 | 700 | Silent Mutation | GAA,GAG | E,E 172 | NP_001269354.1 | |
NM_006572.5 | 700 | Silent Mutation | GAA,GAG | E,E 267 | NP_006563.2 | |
XM_011524202.1 | 700 | Intron | XP_011522504.1 |