Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCATTTTACTCTACCTTTTCATTT[A/G]ATGAAACACCTTTGCATTTTATCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605134 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PITPNC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PITPNC1 - phosphatidylinositol transfer protein, cytoplasmic 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012417.3 | Intron | NP_036549.2 | ||||
NM_181671.2 | Intron | NP_858057.1 | ||||
XM_005257216.2 | Intron | XP_005257273.1 | ||||
XM_017024442.1 | Intron | XP_016879931.1 | ||||
XM_017024443.1 | Intron | XP_016879932.1 | ||||
XM_017024444.1 | Intron | XP_016879933.1 | ||||
XM_017024445.1 | Intron | XP_016879934.1 |