Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGCACCCGCCGGGAGCTGTGCGG[A/G]CGGGGGCGTCCAGGAGAGCGCTGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 142966 MIM: 142960 MIM: 142961 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HOXB-AS3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HOXB-AS3 - HOXB cluster antisense RNA 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HOXB3 - homeobox B3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HOXB5 - homeobox B5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HOXB6 - homeobox B6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018952.4 | 1399 | UTR 3 | NP_061825.2 | |||
XM_005257283.3 | 1399 | UTR 3 | XP_005257340.1 | |||
XM_005257284.3 | 1399 | UTR 3 | XP_005257341.1 | |||
XM_011524727.2 | 1399 | UTR 3 | XP_011523029.1 |