Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTGCCCTCCCATCCGTGTCTTTC[C/T]CTCTTATACATCTCACATACACCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604587 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CALCOCO2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CALCOCO2 - calcium binding and coiled-coil domain 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001261390.1 | Intron | NP_001248319.1 | ||||
NM_001261391.1 | Intron | NP_001248320.1 | ||||
NM_001261393.1 | Intron | NP_001248322.1 | ||||
NM_001261395.1 | Intron | NP_001248324.1 | ||||
NM_005831.4 | Intron | NP_005822.1 |