Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACCCCACCCATCACATAGATGTA[G/T]CACCTTTGGTATAAAATGGGGAGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 120150 MIM: 600119 | ||||||||||||||||||||
Literature Links: |
COL1A1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
COL1A1 - collagen type I alpha 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000088.3 | 5217 | UTR 3 | NP_000079.2 | |||
XM_005257058.3 | 5217 | UTR 3 | XP_005257115.2 | |||
XM_005257059.4 | 5217 | UTR 3 | XP_005257116.2 | |||
XM_011524341.1 | 5217 | UTR 3 | XP_011522643.1 |
SGCA - sarcoglycan alpha | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |