Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCAGATTTTCTTGTCAGTTTCTGA[A/G]AAACCTGGCAGCCTGCTGTTAACAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611519 MIM: 191161 | ||||||||||||||||||||
Literature Links: |
POLDIP2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
POLDIP2 - DNA polymerase delta interacting protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001290145.1 | 3686 | UTR 3 | NP_001277074.1 | |||
NM_015584.4 | 3686 | UTR 3 | NP_056399.1 |
TNFAIP1 - TNF alpha induced protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021137.4 | 3686 | UTR 3 | NP_066960.1 | |||
XM_017024993.1 | 3686 | UTR 3 | XP_016880482.1 |